Product Details

SNP ID
rs116473927
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:154162539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTGTACCTATCAGATAACTTGG[G/T]GTTACTACATCTTATTCCTTATCTC
Phenotype
MIM: 191030
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NUP210L PubMed Links

Gene Details

Gene
NUP210L
Gene Name
nucleoporin 210 like
There are no transcripts associated with this gene.

Gene
TPM3
Gene Name
tropomyosin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001043351.1 6373 Intron NP_001036816.1
NM_001043352.1 6373 Intron NP_001036817.1
NM_001043353.1 6373 Intron NP_001036818.1
NM_001278188.1 6373 Intron NP_001265117.1
NM_001278189.1 6373 Intron NP_001265118.1
NM_001278190.1 6373 Intron NP_001265119.1
NM_001278191.1 6373 Intron NP_001265120.1
NM_152263.3 6373 UTR 3 NP_689476.2
NM_153649.3 6373 Intron NP_705935.1

View Full Product Details