Product Details

SNP ID
rs116484376
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:52692606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCTGAACTCCTACCTCTCCAT[C/T]TTCCTACAATGCACTTTCCCAATGC
Phenotype
MIM: 615623
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COA7 PubMed Links

Gene Details

Gene
COA7
Gene Name
cytochrome c oxidase assembly factor 7 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023077.2 Intron NP_075565.2

View Full Product Details