Product Details

SNP ID
rs116683698
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:158291176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTCCATGTCATCCAGATCTTCT[C/T]ATTTGTCAACCAATCCTGGGCACGA
Phenotype
MIM: 188360 MIM: 188340
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD1B PubMed Links

Gene Details

Gene
CD1B
Gene Name
CD1b molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001764.2 396 Intron NP_001755.1
XM_011510119.2 396 Intron XP_011508421.1
XM_017002784.1 396 Intron XP_016858273.1
XM_017002785.1 396 Intron XP_016858274.1
XM_017002786.1 396 Intron XP_016858275.1
Gene
CD1C
Gene Name
CD1c molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001765.2 396 Missense Mutation TCA,TTA S35L NP_001756.2
XM_005245579.4 396 Missense Mutation TCA,TTA S35L XP_005245636.1

View Full Product Details