Product Details

SNP ID
rs116730493
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:211578122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCCGGGGCGGGTGCTCTTAACG[A/C]GAGGCCCCTTGGGGAGGCCGTGGCC
Phenotype
MIM: 609521
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC30A1 PubMed Links

Gene Details

Gene
SLC30A1
Gene Name
solute carrier family 30 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021194.2 636 Missense Mutation CGC,CTC R164L NP_067017.2

View Full Product Details