Product Details

SNP ID
rs117083668
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:40623602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCATTTGGAGCCGGGACACTGAA[C/G]ATGGCAGTGAGTTGGCCCCAGGGTG
Phenotype
MIM: 611600
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RIMS3 PubMed Links

Gene Details

Gene
RIMS3
Gene Name
regulating synaptic membrane exocytosis 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014747.2 3949 UTR 3 NP_055562.2
XM_011542479.2 3949 UTR 3 XP_011540781.1

View Full Product Details