Product Details

SNP ID
rs117404602
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:247672270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAATGTATAGCTGGAAGCAGGGC[A/G]GATATAGGTGTAGATTACAGGAGAA
Phenotype
MIM: 611677
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR13G1 PubMed Links

Gene Details

Gene
OR13G1
Gene Name
olfactory receptor family 13 subfamily G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005487.1 772 Missense Mutation CGC,TGC R258C NP_001005487.1

View Full Product Details