Product Details

SNP ID
rs118093358
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:214351024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAGGGTAGGTAAAGAAAAGAAAA[A/G]GTGTACTGTTGAGAGTTGATCCATA
Phenotype
MIM: 603155
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTPN14 PubMed Links

Gene Details

Gene
PTPN14
Gene Name
protein tyrosine phosphatase, non-receptor type 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005401.4 11115 UTR 3 NP_005392.2
XM_017001941.1 11115 Intron XP_016857430.1

View Full Product Details