Product Details

SNP ID
rs118126459
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:153438635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGAGAGATAGAGGCCAACAACA[C/T]GTGGAAGAACTAAACCCTGAGCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
S100A7L2 PubMed Links

Gene Details

Gene
S100A7L2
Gene Name
S100 calcium binding protein A7 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001045479.1 Intron NP_001038944.2

View Full Product Details