Product Details

SNP ID
rs121918209
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:173826692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTTTTTTTTTTAAAGAATTC[A/G]GTAGCTTTGTTGTCCGGACCAACAC
Phenotype
MIM: 611503 MIM: 610956
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CENPL PubMed Links

Gene Details

Gene
CENPL
Gene Name
centromere protein L
There are no transcripts associated with this gene.

Gene
DARS2
Gene Name
aspartyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018122.4 704 Missense Mutation AGT,GGT S45G NP_060592.2
XM_006711427.3 704 Missense Mutation AGT,GGT S45G XP_006711490.1
XM_011509711.2 704 Missense Mutation AGT,GGT S45G XP_011508013.1

View Full Product Details