Product Details

SNP ID
rs137925587
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:248239277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGCTGAATGTTTCTTGTTGGCT[C/G]TCATGGCTTATGACCGCTATGTGGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR2M4 PubMed Links

Gene Details

Gene
OR2M4
Gene Name
olfactory receptor family 2 subfamily M member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017504.1 349 Missense Mutation CTC,GTC L117V NP_059974.1

View Full Product Details