Product Details
- SNP ID
-
rs138121177
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:150997633 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGCAGCAGCCCACCTGCAGTGA[C/T]AGGACCCGCGTCCGCATCCGCACTG
- Phenotype
-
MIM: 603319
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA9
PubMed Links
Gene Details
- Gene
- ANXA9
- Gene Name
- annexin A9
There are no transcripts associated with this gene.
- Gene
- FAM63A
- Gene Name
- family with sequence similarity 63 member A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001040217.2 |
2000 |
Silent Mutation |
CTA,CTG |
L298L |
NP_001035307.1 |
NM_001163258.1 |
2000 |
Silent Mutation |
CTA,CTG |
L488L |
NP_001156730.1 |
NM_001163259.1 |
2000 |
Silent Mutation |
CTA,CTG |
L345L |
NP_001156731.1 |
NM_001163260.1 |
2000 |
Silent Mutation |
CTA,CTG |
L298L |
NP_001156732.1 |
NM_001319998.1 |
2000 |
Silent Mutation |
CTA,CTG |
L440L |
NP_001306927.1 |
NM_018379.4 |
2000 |
Silent Mutation |
CTA,CTG |
L440L |
NP_060849.2 |
XM_005245321.3 |
2000 |
Silent Mutation |
CTA,CTG |
L441L |
XP_005245378.1 |
XM_005245325.3 |
2000 |
Silent Mutation |
CTA,CTG |
L299L |
XP_005245382.1 |
XM_005245327.3 |
2000 |
Silent Mutation |
CTA,CTG |
L275L |
XP_005245384.1 |
XM_005245328.3 |
2000 |
Silent Mutation |
CTA,CTG |
L274L |
XP_005245385.1 |
XM_011509757.2 |
2000 |
Silent Mutation |
CTA,CTG |
L432L |
XP_011508059.1 |
XM_011509759.2 |
2000 |
Silent Mutation |
CTA,CTG |
L387L |
XP_011508061.1 |
XM_017001770.1 |
2000 |
Silent Mutation |
CTA,CTG |
L440L |
XP_016857259.1 |
XM_017001771.1 |
2000 |
Silent Mutation |
CTA,CTG |
L440L |
XP_016857260.1 |
XM_017001772.1 |
2000 |
Silent Mutation |
CTA,CTG |
L440L |
XP_016857261.1 |
XM_017001773.1 |
2000 |
Silent Mutation |
CTA,CTG |
L431L |
XP_016857262.1 |
XM_017001774.1 |
2000 |
Silent Mutation |
CTA,CTG |
L431L |
XP_016857263.1 |
XM_017001775.1 |
2000 |
Silent Mutation |
CTA,CTG |
L387L |
XP_016857264.1 |
XM_017001776.1 |
2000 |
Silent Mutation |
CTA,CTG |
L378L |
XP_016857265.1 |
XM_017001777.1 |
2000 |
Silent Mutation |
CTA,CTG |
L488L |
XP_016857266.1 |
View Full Product Details