Product Details
- SNP ID
-
rs138204543
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:19076822 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTCAGGGCTTTGTCGGCAGCTTC[A/G]TAGATGGGCATGTCGTTGTGGCGGA
- Phenotype
-
MIM: 609890
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UBR4
PubMed Links
Gene Details
- Gene
- UBR4
- Gene Name
- ubiquitin protein ligase E3 component n-recognin 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020765.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5135Y |
NP_065816.2 |
XM_011541108.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5186Y |
XP_011539410.1 |
XM_011541109.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5185Y |
XP_011539411.1 |
XM_011541110.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5185Y |
XP_011539412.1 |
XM_011541111.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5185Y |
XP_011539413.1 |
XM_011541112.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5181Y |
XP_011539414.1 |
XM_011541113.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5180Y |
XP_011539415.1 |
XM_011541114.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5180Y |
XP_011539416.1 |
XM_011541115.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5178Y |
XP_011539417.1 |
XM_011541116.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5175Y |
XP_011539418.1 |
XM_011541117.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5158Y |
XP_011539419.1 |
XM_011541118.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5157Y |
XP_011539420.1 |
XM_011541119.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5146Y |
XP_011539421.1 |
XM_011541120.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5145Y |
XP_011539422.1 |
XM_011541121.2 |
15386 |
Silent Mutation |
TAC,TAT |
Y5134Y |
XP_011539423.1 |
XM_017000822.1 |
15386 |
Silent Mutation |
TAC,TAT |
Y5179Y |
XP_016856311.1 |
XM_017000823.1 |
15386 |
Silent Mutation |
TAC,TAT |
Y5170Y |
XP_016856312.1 |
XM_017000824.1 |
15386 |
Silent Mutation |
TAC,TAT |
Y5152Y |
XP_016856313.1 |
XM_017000825.1 |
15386 |
Silent Mutation |
TAC,TAT |
Y5147Y |
XP_016856314.1 |
XM_017000826.1 |
15386 |
Silent Mutation |
TAC,TAT |
Y5146Y |
XP_016856315.1 |
XM_017000827.1 |
15386 |
Silent Mutation |
TAC,TAT |
Y5141Y |
XP_016856316.1 |
XM_017000828.1 |
15386 |
Silent Mutation |
TAC,TAT |
Y5133Y |
XP_016856317.1 |
XM_017000829.1 |
15386 |
Silent Mutation |
TAC,TAT |
Y5117Y |
XP_016856318.1 |
XM_017000830.1 |
15386 |
Silent Mutation |
TAC,TAT |
Y5100Y |
XP_016856319.1 |
View Full Product Details