Product Details

SNP ID
rs138398618
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:154569241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTTAGTCTCAAGTCATCTCTGT[A/G]GAGCTGTGTAGACTCTTTGGTCAGT
Phenotype
MIM: 118507
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRNB2 PubMed Links

Gene Details

Gene
CHRNB2
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000748.2 Intron NP_000739.1
XM_017000180.1 Intron XP_016855669.1

View Full Product Details