Product Details

SNP ID
rs138405719
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:248811267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGCCCTGTAAGTTCTGCTTTGG[A/G]GCACACATCGCAGAGTGCCTCAGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SH3BP5L PubMed Links

Gene Details

Gene
SH3BP5L
Gene Name
SH3 binding domain protein 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322462.1 2255 UTR 3 NP_001309391.1
NM_001322463.1 2255 UTR 3 NP_001309392.1
NM_001322464.1 2255 UTR 3 NP_001309393.1
NM_030645.1 2255 UTR 3 NP_085148.1
XM_017002404.1 2255 UTR 3 XP_016857893.1
XM_017002405.1 2255 UTR 3 XP_016857894.1
XM_017002406.1 2255 UTR 3 XP_016857895.1
XM_017002407.1 2255 UTR 3 XP_016857896.1

View Full Product Details