Product Details
- SNP ID
-
rs138440339
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:109690472 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGCAGATCACTTTTGTAGATTTT[C/G]TCGTCTATGATGTCCTTGACCTCCA
- Phenotype
-
MIM: 138350
MIM: 138380
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GSTM1
PubMed Links
- Additional Information
-
The GSTM1 gene exhibits copy number variation. Individuals may carry GSTM1 deletion alleles. GSTM1 SNP genotyping assays run samples having no GSTM1 genes will not amplify, and samples having 1 gene copy will cluster with one of the homozygous clusters. For accurate GSTM1 genotype analysis, copy number analysis must be done. For more information, refer to the PGx Experiments User Guide (Pub. # MAN0009612) Chapter 2 Copy Number Variation section.
Gene Details
- Gene
- GSTM1
- Gene Name
- glutathione S-transferase mu 1
- Gene
- GSTM2
- Gene Name
- glutathione S-transferase mu 2 (muscle)
There are no transcripts associated with this gene.
View Full Product Details