Product Details

SNP ID
rs138571349
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:13476199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTACAAAGCCAGCAATGTGCAAG[C/T]ATCCTCCCAAAAATAAAAGGGGGGA
Phenotype
MIM: 615212
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC38 PubMed Links

Gene Details

Gene
LRRC38
Gene Name
leucine rich repeat containing 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010847.1 Intron NP_001010847.1

View Full Product Details