Product Details
- SNP ID
-
rs138628609
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:1312072 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTGCCTGGGTCCTCAGAAGGGGC[A/G]GCGGCCTGGAGGAGGACGGACTCCA
- Phenotype
-
MIM: 611354
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACAP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12103] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACAP3
- Gene Name
- ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
There are no transcripts associated with this gene.
- Gene
- CPSF3L
- Gene Name
- cleavage and polyadenylation specific factor 3-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256456.1 |
1917 |
Silent Mutation |
GCC,GCT |
A567A |
NP_001243385.1 |
NM_001256460.1 |
1917 |
Silent Mutation |
GCC,GCT |
A532A |
NP_001243389.1 |
NM_001256462.1 |
1917 |
Silent Mutation |
GCC,GCT |
A463A |
NP_001243391.1 |
NM_001256463.1 |
1917 |
Silent Mutation |
GCC,GCT |
A460A |
NP_001243392.1 |
NM_017871.5 |
1917 |
Silent Mutation |
GCC,GCT |
A561A |
NP_060341.2 |
XM_011541647.1 |
1917 |
Silent Mutation |
GCC,GCT |
A621A |
XP_011539949.1 |
XM_011541648.1 |
1917 |
Silent Mutation |
GCC,GCT |
A583A |
XP_011539950.1 |
XM_011541650.1 |
1917 |
Silent Mutation |
GCC,GCT |
A437A |
XP_011539952.1 |
XM_017001557.1 |
1917 |
Silent Mutation |
GCC,GCT |
A437A |
XP_016857046.1 |
XM_017001558.1 |
1917 |
Silent Mutation |
GCC,GCT |
A437A |
XP_016857047.1 |
- Gene
- MIR6727
- Gene Name
- microRNA 6727
There are no transcripts associated with this gene.
- Gene
- PUSL1
- Gene Name
- pseudouridylate synthase-like 1
View Full Product Details