Product Details

SNP ID
rs138668107
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:179592766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACCACTCCGAATCCTTGGGTAT[C/T]GGTCCACTATGGAGCTGGTATTGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TDRD5 PubMed Links

Gene Details

Gene
TDRD5
Gene Name
tudor domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199085.1 669 Missense Mutation CGG,TGG R51W NP_001186014.1
NM_001199089.1 669 Missense Mutation CGG,TGG R51W NP_001186018.1
NM_001199091.1 669 Missense Mutation CGG,TGG R51W NP_001186020.1
NM_001199092.1 669 Intron NP_001186021.1
NM_173533.3 669 Missense Mutation CGG,TGG R51W NP_775804.2
XM_005244934.1 669 Missense Mutation CGG,TGG R51W XP_005244991.1
XM_005244935.4 669 Missense Mutation CGG,TGG R51W XP_005244992.1
XM_017000473.1 669 Missense Mutation CGG,TGG R51W XP_016855962.1

View Full Product Details