Product Details

SNP ID
rs138691534
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:153330832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCCTGCCCAGGAGACAAGGTTC[A/G]GGCCACATCACTGTGGCCCACCAGC
Phenotype
MIM: 608198
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGLYRP4 PubMed Links

Gene Details

Gene
PGLYRP4
Gene Name
peptidoglycan recognition protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020393.3 1417 Nonsense Mutation CGA,TGA R353* NP_065126.2
XM_011509789.2 1417 Nonsense Mutation CGA,TGA R353* XP_011508091.1
XM_011509790.1 1417 Nonsense Mutation CGA,TGA R353* XP_011508092.1
XM_011509791.2 1417 Nonsense Mutation CGA,TGA R349* XP_011508093.1
XM_011509792.1 1417 Intron XP_011508094.1
XM_011509793.2 1417 Nonsense Mutation CGA,TGA R185* XP_011508095.1

View Full Product Details