Product Details

SNP ID
rs138756182
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:112701679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGCCAAAGGCACTGATCCGGTTC[A/G]CCATGTCCCGGCCTTCCTCAGACCG
Phenotype
MIM: 610742 MIM: 609957 MIM: 165380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MOV10 PubMed Links

Gene Details

Gene
MOV10
Gene Name
Mov10 RISC complex RNA helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130079.2 4634 Intron NP_001123551.1
NM_001286072.1 4634 Intron NP_001273001.1
NM_001321324.1 4634 Intron NP_001308253.1
NM_020963.4 4634 Intron NP_066014.1
XM_005270869.4 4634 UTR 3 XP_005270926.1
XM_005270870.4 4634 UTR 3 XP_005270927.1
XM_017001318.1 4634 UTR 3 XP_016856807.1
XM_017001319.1 4634 Intron XP_016856808.1
Gene
PPM1J
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1J
There are no transcripts associated with this gene.

Gene
RHOC
Gene Name
ras homolog family member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042678.1 4634 Missense Mutation GCG,GTG A148V NP_001036143.1
NM_001042679.1 4634 Missense Mutation GCG,GTG A148V NP_001036144.1
NM_175744.4 4634 Missense Mutation GCG,GTG A148V NP_786886.1

View Full Product Details