Product Details

SNP ID
rs138793608
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:35560716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATGCCAGGCATACCTGGGGCAC[A/G]GCTATGGCTTTGACCAGGCCCTGGC
Phenotype
MIM: 613535 MIM: 608458
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0319L PubMed Links

Gene Details

Gene
KIAA0319L
Gene Name
KIAA0319 like
There are no transcripts associated with this gene.

Gene
NCDN
Gene Name
neurochondrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014839.1 624 Missense Mutation AGC,GGC S189G NP_001014839.1
NM_001014841.1 624 Missense Mutation AGC,GGC S172G NP_001014841.1
NM_014284.2 624 Missense Mutation AGC,GGC S189G NP_055099.1

View Full Product Details