Product Details
- SNP ID
-
rs138799860
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:92513689 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTTGAATACGACTCTCTTCTTCT[C/T]GGGGGCCGCTCCTTTTGAACCACTT
- Phenotype
-
MIM: 602942
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EVI5
PubMed Links
Gene Details
- Gene
- EVI5
- Gene Name
- ecotropic viral integration site 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001308248.1 |
2522 |
Silent Mutation |
CCA,CCG |
P811P |
NP_001295177.1 |
NM_005665.5 |
2522 |
Silent Mutation |
CCA,CCG |
P800P |
NP_005656.4 |
XM_017002269.1 |
2522 |
Silent Mutation |
CCA,CCG |
P920P |
XP_016857758.1 |
XM_017002270.1 |
2522 |
Silent Mutation |
CCA,CCG |
P917P |
XP_016857759.1 |
XM_017002271.1 |
2522 |
Silent Mutation |
CCA,CCG |
P893P |
XP_016857760.1 |
XM_017002272.1 |
2522 |
Silent Mutation |
CCA,CCG |
P887P |
XP_016857761.1 |
XM_017002273.1 |
2522 |
Silent Mutation |
CCA,CCG |
P876P |
XP_016857762.1 |
XM_017002274.1 |
2522 |
Silent Mutation |
CCA,CCG |
P876P |
XP_016857763.1 |
XM_017002275.1 |
2522 |
Silent Mutation |
CCA,CCG |
P876P |
XP_016857764.1 |
XM_017002276.1 |
2522 |
Silent Mutation |
CCA,CCG |
P849P |
XP_016857765.1 |
XM_017002277.1 |
2522 |
Silent Mutation |
CCA,CCG |
P844P |
XP_016857766.1 |
XM_017002278.1 |
2522 |
Silent Mutation |
CCA,CCG |
P838P |
XP_016857767.1 |
XM_017002279.1 |
2522 |
Silent Mutation |
CCA,CCG |
P831P |
XP_016857768.1 |
XM_017002280.1 |
2522 |
Silent Mutation |
CCA,CCG |
P816P |
XP_016857769.1 |
XM_017002281.1 |
2522 |
Silent Mutation |
CCA,CCG |
P808P |
XP_016857770.1 |
XM_017002282.1 |
2522 |
UTR 3 |
|
|
XP_016857771.1 |
XM_017002283.1 |
2522 |
UTR 3 |
|
|
XP_016857772.1 |
XM_017002284.1 |
2522 |
Silent Mutation |
CCA,CCG |
P767P |
XP_016857773.1 |
XM_017002285.1 |
2522 |
UTR 3 |
|
|
XP_016857774.1 |
XM_017002286.1 |
2522 |
Silent Mutation |
CCA,CCG |
P712P |
XP_016857775.1 |
XM_017002287.1 |
2522 |
Silent Mutation |
CCA,CCG |
P712P |
XP_016857776.1 |
XM_017002288.1 |
2522 |
Silent Mutation |
CCA,CCG |
P712P |
XP_016857777.1 |
View Full Product Details