Product Details

SNP ID
rs138906127
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:226361468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGCTACCTCTCCCAATTACCACA[G/T]GGAGGTCTTAAAATTGAATTTCAGT
Phenotype
MIM: 173870
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PARP1 PubMed Links

Gene Details

Gene
PARP1
Gene Name
poly(ADP-ribose) polymerase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001618.3 3208 Missense Mutation ATG,CTG M1013L NP_001609.2

View Full Product Details