Product Details

SNP ID
rs138949224
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:230323469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCAAGCCCAGCAGCTCTCTGA[C/T]GAGTCTCTCTCCAAACTGCGCCCGG
Phenotype
MIM: 616791
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGBD5 PubMed Links

Gene Details

Gene
PGBD5
Gene Name
piggyBac transposable element derived 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258311.1 2600 Missense Mutation ATC,GTC I511V NP_001245240.1
XM_017002330.1 2600 Missense Mutation ATC,GTC I422V XP_016857819.1

View Full Product Details