Product Details

SNP ID
rs139067472
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:28334872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGGAATTAAGTGTGGTAGCACCC[A/G]CTGGGCAGGACATGGTCTCTGATGA
Phenotype
MIM: 612384
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED18 PubMed Links

Gene Details

Gene
MED18
Gene Name
mediator complex subunit 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127350.1 Intron NP_001120822.1
NM_017638.2 Intron NP_060108.2
XM_005245914.4 Intron XP_005245971.1

View Full Product Details