Product Details

SNP ID
rs139088461
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:54063192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAATGTGATGTGGGGAGCTAGAA[C/T]TGATCTCCTATGTGAAGCTGGAAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4781 PubMed Links

Gene Details

Gene
MIR4781
Gene Name
microRNA 4781
There are no transcripts associated with this gene.

Gene
TCEANC2
Gene Name
transcription elongation factor A N-terminal and central domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153035.2 Intron NP_694580.1
XM_017000293.1 Intron XP_016855782.1
Gene
TMEM59
Gene Name
transmembrane protein 59
There are no transcripts associated with this gene.

View Full Product Details