Product Details

SNP ID
rs139249390
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:28603573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGTTTTCCGGATCAATGCCATT[C/T]TCTGAAAGGAGAGACAAATAAGCAG
Phenotype
MIM: 600773
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB42 PubMed Links

Gene Details

Gene
RAB42
Gene Name
RAB42, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
TAF12
Gene Name
TATA-box binding protein associated factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135218.1 637 Missense Mutation AAA,AGA K151R NP_001128690.1
NM_005644.3 637 Missense Mutation AAA,AGA K151R NP_005635.1
XM_017002186.1 637 Missense Mutation AAA,AGA K231R XP_016857675.1

View Full Product Details