Product Details

SNP ID
rs139561979
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:37475598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCCGTCAGGGCACCCCAAGGCC[A/G]GGTCAAGAGCTGGCCGCTGAGGAGG
Phenotype
MIM: 610562
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01137 PubMed Links

Gene Details

Gene
LINC01137
Gene Name
long intergenic non-protein coding RNA 1137
There are no transcripts associated with this gene.

Gene
MIR6732
Gene Name
microRNA 6732
There are no transcripts associated with this gene.

Gene
ZC3H12A
Gene Name
zinc finger CCCH-type containing 12A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323550.1 252 Silent Mutation CCA,CCG P34P NP_001310479.1
NM_001323551.1 252 UTR 5 NP_001310480.1
NM_025079.2 252 Silent Mutation CCA,CCG P34P NP_079355.2
XM_011542198.2 252 Silent Mutation CCA,CCG P34P XP_011540500.1

View Full Product Details