Product Details

SNP ID
rs139780555
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:155013511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGCAGAGCCAGGATTTAAAGCAG[A/G]TCTCACCCCCTTCCAGGCCCCAACC
Phenotype
MIM: 607646
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCST2 PubMed Links

Gene Details

Gene
DCST2
Gene Name
DC-STAMP domain containing 2
There are no transcripts associated with this gene.

Gene
ZBTB7B
Gene Name
zinc finger and BTB domain containing 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252406.2 Intron NP_001239335.1
NM_001256455.1 Intron NP_001243384.1
XM_006711349.2 Intron XP_006711412.2
XM_006711353.1 Intron XP_006711416.1
XM_006711354.1 Intron XP_006711417.1
XM_006711356.3 Intron XP_006711419.1
XM_006711357.1 Intron XP_006711420.1
XM_006711358.1 Intron XP_006711421.1
XM_006711359.2 Intron XP_006711422.1
XM_011509598.2 Intron XP_011507900.1
XM_011509599.2 Intron XP_011507901.1
XM_017001399.1 Intron XP_016856888.1

View Full Product Details