Product Details

SNP ID
rs139847791
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:152085232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAACTTGCTTTTGATGCTCCTCT[C/T]GGGTTACATTTGTTTGCTGCAGTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100131107 PubMed Links

Gene Details

Gene
LOC100131107
Gene Name
putative UPF0607 protein ENSP00000383783
There are no transcripts associated with this gene.

Gene
TCHHL1
Gene Name
trichohyalin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008536.1 2515 Missense Mutation CAA,CGA Q817R NP_001008536.1

View Full Product Details