Product Details

SNP ID
rs139927527
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:197088421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTACATCATAAACTGCTGAAGT[A/G]GTTTTCTCATACTTGAAGAGAACAG
Phenotype
MIM: 605481
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASPM PubMed Links

Gene Details

Gene
ASPM
Gene Name
abnormal spindle microtubule assembly
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206846.1 10253 Silent Mutation ACC,ACT T1747T NP_001193775.1
NM_018136.4 10253 Silent Mutation ACC,ACT T3332T NP_060606.3

View Full Product Details