Product Details

SNP ID
rs140125203
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:203496234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGATTATGGGGACCAACTCCCC[A/G]AGGTGAGGGACACAGCAGACCAACT
Phenotype
MIM: 605127 MIM: 601914
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OPTC PubMed Links

Gene Details

Gene
OPTC
Gene Name
opticin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014359.3 345 Missense Mutation AAG,GAG K77E NP_055174.1
XM_011509406.2 345 Missense Mutation AAG,GAG K77E XP_011507708.1
Gene
PRELP
Gene Name
proline and arginine rich end leucine rich repeat protein
There are no transcripts associated with this gene.

View Full Product Details