Product Details

SNP ID
rs140339536
Assay Type
Functionally Tested
NCBI dbSNP Submissions
5
Location
Chr.1:1243120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACACACACCACGTCCCGGGCCC[A/G]CAGCCGGGCCTTGCCCTGCAGCTCA
Phenotype
MIM: 615291 MIM: 616593
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
B3GALT6 PubMed Links

Gene Details

Gene
B3GALT6
Gene Name
beta-1,3-galactosyltransferase 6
There are no transcripts associated with this gene.

Gene
FAM132A
Gene Name
family with sequence similarity 132 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014980.2 799 Missense Mutation CGG,TGG R225W NP_001014980.1
XM_011541435.2 799 Silent Mutation TGC,TGT C229C XP_011539737.1
XM_011541436.2 799 Silent Mutation TGC,TGT C223C XP_011539738.1
XM_011541437.1 799 Silent Mutation TGC,TGT C157C XP_011539739.1

View Full Product Details