Product Details

SNP ID
rs140341596
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:87331207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTTCAATGATCTATTATTCTTG[C/T]CTGTGTCCTGAAAACCCAGAGAAAA
Phenotype
MIM: 603129
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LMO4 PubMed Links

Gene Details

Gene
LMO4
Gene Name
LIM domain only 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006769.3 2340 Intron NP_006760.1
XM_005271291.3 2340 UTR 5 XP_005271348.1

View Full Product Details