Product Details

SNP ID
rs140555845
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:179593750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACATGCAATATGGATTTCTCTCT[A/C]TGTTTGAAGTGCTTAATGCGGCTTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TDRD5 PubMed Links

Gene Details

Gene
TDRD5
Gene Name
tudor domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199085.1 1041 Missense Mutation ATG,CTG M175L NP_001186014.1
NM_001199089.1 1041 Missense Mutation ATG,CTG M175L NP_001186018.1
NM_001199091.1 1041 Missense Mutation ATG,CTG M175L NP_001186020.1
NM_001199092.1 1041 Intron NP_001186021.1
NM_173533.3 1041 Missense Mutation ATG,CTG M175L NP_775804.2
XM_005244934.1 1041 Missense Mutation ATG,CTG M175L XP_005244991.1
XM_005244935.4 1041 Missense Mutation ATG,CTG M175L XP_005244992.1
XM_017000473.1 1041 Missense Mutation ATG,CTG M175L XP_016855962.1

View Full Product Details