Product Details

SNP ID
rs140561898
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:204201687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAACATGCCGAGGAGGGGCCAG[C/T]GTAGGAGCACGATAACCACACCCCC
Phenotype
MIM: 603286
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOLT1A PubMed Links

Gene Details

Gene
GOLT1A
Gene Name
golgi transport 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198447.1 428 Missense Mutation CAC,CGC H81R NP_940849.1
XM_017000314.1 428 Missense Mutation CAC,CGC H81R XP_016855803.1
Gene
KISS1
Gene Name
KiSS-1 metastasis-suppressor
There are no transcripts associated with this gene.

View Full Product Details