Product Details
- SNP ID
-
rs140566193
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:943917 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCATCTCTCTGCAGGTGGCCAGG[C/T]GCCTGGGCCGAGTTTTCTACGTGGC
- Phenotype
-
MIM: 610770
MIM: 616765
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NOC2L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115454328,rs7523549] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NOC2L
- Gene Name
- NOC2 like nucleolar associated transcriptional repressor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015658.3 |
1890 |
Intron |
|
|
NP_056473.2 |
- Gene
- SAMD11
- Gene Name
- sterile alpha motif domain containing 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152486.2 |
1890 |
Missense Mutation |
CGC,TGC |
R604C |
NP_689699.2 |
View Full Product Details