Product Details

SNP ID
rs140898399
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:42154830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCAGGACCTTCAGCCTGTCTGC[A/G]CCTCGCAGGAGGCTTCCAGCATCTT
Phenotype
MIM: 139392 MIM: 601271
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GUCA2A PubMed Links

Gene Details

Gene
GUCA2A
Gene Name
guanylate cyclase activator 2A
There are no transcripts associated with this gene.

Gene
GUCA2B
Gene Name
guanylate cyclase activator 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007102.2 271 Missense Mutation ACC,GCC T81A NP_009033.1

View Full Product Details