Product Details

SNP ID
rs140904380
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:158255122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCCTTCCATGTCACCTGGATC[A/G]CATCCTTTTACAACCATTCCTGGAA
Phenotype
MIM: 188370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD1A PubMed Links

Gene Details

Gene
CD1A
Gene Name
CD1a molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320652.1 122 Missense Mutation ACA,GCA T22A NP_001307581.1
NM_001763.2 122 Missense Mutation ACA,GCA T33A NP_001754.2

View Full Product Details