Product Details

SNP ID
rs141077306
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:26474131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCCAAGCCTAAAAAGGCCCCTG[C/T]AAAGGTAAGTGCTAACATTGGAACT
Phenotype
MIM: 608172 MIM: 163910
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHDDS PubMed Links

Gene Details

Gene
DHDDS
Gene Name
dehydrodolichyl diphosphate synthase subunit
There are no transcripts associated with this gene.

Gene
HMGN2
Gene Name
high mobility group nucleosomal binding domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005517.3 339 Missense Mutation GCA,GTA A46V NP_005508.1
Gene
LOC101928324
Gene Name
uncharacterized LOC101928324
There are no transcripts associated with this gene.

View Full Product Details