Product Details

SNP ID
rs141100657
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:156700982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTTTGATGTAGAAAGTGTCTCC[C/T]TCCTGTTTGATCTCCACTGCTGGCT
Phenotype
MIM: 180231
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRABP2 PubMed Links

Gene Details

Gene
CRABP2
Gene Name
cellular retinoic acid binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199723.1 362 Silent Mutation GAA,GAG E47E NP_001186652.1
NM_001878.3 362 Silent Mutation GAA,GAG E47E NP_001869.1
XM_017000343.1 362 Silent Mutation GAA,GAG E47E XP_016855832.1

View Full Product Details