Product Details

SNP ID
rs141225393
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:36137826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGCTCCCCGCAACACCCCACA[A/C]AAGAGATTGGAATAAATAAGGGATG
Phenotype
MIM: 610955
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRAPPC3 PubMed Links

Gene Details

Gene
TRAPPC3
Gene Name
trafficking protein particle complex 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270894.1 331 Missense Mutation TTG,TTT L139F NP_001257823.1
NM_001270895.1 331 Missense Mutation TTG,TTT L85F NP_001257824.1
NM_001270896.1 331 Missense Mutation TTG,TTT L85F NP_001257825.1
NM_001270897.1 331 Missense Mutation TTG,TTT L65F NP_001257826.1
NM_014408.4 331 Missense Mutation TTG,TTT L131F NP_055223.1

View Full Product Details