Product Details

SNP ID
rs141236032
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:89132320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCTCTTCATTTTCAGCTGCTTC[A/G]ATTTGTTCTTTCAGTCGATTAATCT
Phenotype
MIM: 600412 MIM: 612468
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GBP2 PubMed Links

Gene Details

Gene
GBP2
Gene Name
guanylate binding protein 2
There are no transcripts associated with this gene.

Gene
GBP7
Gene Name
guanylate binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207398.2 1885 Silent Mutation ATC,ATT I582I NP_997281.2

View Full Product Details