Product Details

SNP ID
hCV166294015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:149929885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCCTCTAGAGAAGAGCCACACA[A/G]AACTGGGGGGTCCAGGAACCATGAA
Phenotype
MIM: 605593
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTMR11 PubMed Links

Gene Details

Gene
MTMR11
Gene Name
myotubularin related protein 11
There are no transcripts associated with this gene.

Gene
SF3B4
Gene Name
splicing factor 3b subunit 4
There are no transcripts associated with this gene.

View Full Product Details