Product Details

SNP ID
rs141314271
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:37569097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAACCAGGTCATCCCCAGAAAAC[C/T]GAGGCACCACGTTGATTTTACCAAA
Phenotype
MIM: 602135 MIM: 609365
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNALI1 PubMed Links

Gene Details

Gene
DNALI1
Gene Name
dynein axonemal light intermediate chain 1
There are no transcripts associated with this gene.

Gene
GNL2
Gene Name
G protein nucleolar 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323623.1 1785 Missense Mutation CAG,CGG Q609R NP_001310552.1
NM_001323624.1 1785 Missense Mutation CAG,CGG Q358R NP_001310553.1
NM_013285.2 1785 Missense Mutation CAG,CGG Q541R NP_037417.1

View Full Product Details