Product Details

SNP ID
rs141349380
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:943354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCTGCAGCTTCGTGGGGGGCCTG[G/T]CTGGCTGTGGAGAGTACACTCGGGT
Phenotype
MIM: 610770 MIM: 616765
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NOC2L PubMed Links

Gene Details

Gene
NOC2L
Gene Name
NOC2 like nucleolar associated transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015658.3 1746 Intron NP_056473.2
Gene
SAMD11
Gene Name
sterile alpha motif domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152486.2 1746 Missense Mutation GCT,TCT A556S NP_689699.2

View Full Product Details