Product Details

SNP ID
rs141402675
Assay Type
Functionally Tested
NCBI dbSNP Submissions
11
Location
Chr.1:197086979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACCTACTTCGTACATCCTACAA[A/T]ATAAAATGCACAGTTACTAAAAAGT
Phenotype
MIM: 605481
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
ASPM PubMed Links

Gene Details

Gene
ASPM
Gene Name
abnormal spindle microtubule assembly
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206846.1 Intron NP_001193775.1
NM_018136.4 Intron NP_060606.3

View Full Product Details