Product Details
- SNP ID
-
hCV166304895
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:145730853 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAGGGGTTTGTAGTGTTTTTGTG[T/C]AAGGACAGGTCTGGAGACTGTTCTC
- Phenotype
-
MIM: 604463
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD160
PubMed Links
Gene Details
- Gene
- CD160
- Gene Name
- CD160 molecule
There are no transcripts associated with this gene.
View Full Product Details