Product Details

SNP ID
rs141455584
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:1916497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACAAGGGAGTCTTTGAGATGTT[C/T]GACGAAGAGGGCAACGGGGAGGTGA
Phenotype
MIM: 610171
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALML6 PubMed Links

Gene Details

Gene
CALML6
Gene Name
calmodulin like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005244729.3 1731 Silent Mutation TTC,TTT F67F XP_005244786.2
XM_011540841.2 1731 Silent Mutation TTC,TTT F28F XP_011539143.1
Gene
CFAP74
Gene Name
cilia and flagella associated protein 74
There are no transcripts associated with this gene.

Gene
TMEM52
Gene Name
transmembrane protein 52
There are no transcripts associated with this gene.

View Full Product Details