Product Details

SNP ID
rs141572839
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:37556931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGAGTGAGGGGGCGGTTTCCAT[A/G]GTGACGGCAAACAAGGCCCACACTG
Phenotype
MIM: 602135 MIM: 609365 MIM: 608241
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNALI1 PubMed Links

Gene Details

Gene
DNALI1
Gene Name
dynein axonemal light intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003462.3 13 Missense Mutation ATA,ATG I1M NP_003453.2
XM_005271172.2 13 Intron XP_005271229.1
Gene
GNL2
Gene Name
G protein nucleolar 2
There are no transcripts associated with this gene.

Gene
SNIP1
Gene Name
Smad nuclear interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details